Assay Details
| Gene Symbol: LOC102124971 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015449993.1 | XP_015305479.1 | 1 - 2 | 972 | 107 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102124971| Gene Symbol: LOC102124971 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015449993.1 | XP_015305479.1 | 1 - 2 | 972 | 107 | |
Set Membership: |
|