Assay Details
| Gene Symbol: RPL37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005595986.2 | XP_005596043.2 | 1 - 2 | 289 | 102 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005595986.2 transcripts.
Set Membership: |
|

RPL37
Genomic Map