Assay Details
| Gene Symbol: LOC102136965 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005590508.2 | XP_005590565.1 | 1 - 2 | 281 | 117 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005590508.2 transcripts.
Set Membership: |
|

LOC102136965
Genomic Map