Assay Details
| Gene Symbol: DPEP2NB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015442780.1 | XP_015298266.1 | 1 | 626 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015442780.1 transcripts.
Set Membership: |
|

DPEP2NB
Genomic Map