Assay Details
| Gene Symbol: LOC102131813 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015453363.1 | XP_015308849.1 | 2 | 1921 | 97 | |
| XM_015453364.1 | XP_015308850.1 | 3 | 469 | 97 | ||
More Information
Set Membership: |
|

LOC102131813
Genomic Map