Assay Details
| Gene Symbol: Syt11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018804.3 | NP_061274.2 | 3 - 4 | 1250 | 68 | |
| GenBank mRNA | AB026808.1 | - | 975 | 68 | ||
| AK122200.1 | - | 2 - 3 | 744 | 68 | ||
| AK144169.1 | - | 3 - 4 | 1168 | 68 | ||
| AK158745.1 | - | 3 - 4 | 1146 | 68 | ||
| AK164082.1 | - | 3 - 4 | 1168 | 68 | ||
| BC025207.1 | - | 2 - 3 | 468 | 5030173 | 68 | |
| BC054526.1 | - | 3 - 4 | 1191 | 5698412 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1956267 | CD776044.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1956267 | NM_018804.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 786383 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 86238 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 869054 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 966260 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 411502 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 375578 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 277611 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 384481 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 788408 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 710108 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 494559 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 267525 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 749707 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 146510 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 351931 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 700071 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 43339 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 25115 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 8180 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P153620 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1956267 | CD776044.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1956267 | NM_018804.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P260730 | BC054526.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P260730 | NM_018804.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P260730 | AK018343.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P260730 | AK122200.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P125935 | BC054526.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P125935 | BC025207.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P125935 | AK158745.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P125935 | AK164082.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P125935 | AK144169.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P125935 | BC054526.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P125935 | BC025207.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P125935 | AK158745.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P125935 | AK164082.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P125935 | AK144169.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P260730 | BC054526.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P260730 | NM_018804.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P260730 | AK018343.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P260730 | AK122200.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P107381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22315453 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22315504 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02562549 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22315508 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22315487 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22315480 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02808285 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22315582 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22315468 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22315478 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22315512 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22315475 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22315506 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22315484 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22315467 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02575814 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22315462 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22315453 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22315486 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22315464 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02398215 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22315469 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22315584 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02808276 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22315500 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22315472 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22315485 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22315477 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22315482 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02594201 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22315466 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22315463 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10499432 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499437 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499433 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499436 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499434 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499435 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449264_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429314_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429315_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1456464_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455176_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10499431 | - | - |
| Mouse Genome 430 2.0 Array | 1436176_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1429314_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1429315_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1455176_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1429314_at | - | - |
| Mouse Genome 430 2.0 Array | 1429315_at | - | - |
| Mouse Genome 430 2.0 Array | 1429729_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1455176_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1436176_at | - | - |
| Mouse Genome 430 2.0 Array | 1456464_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1449264_at | - | - |
| Mouse Genome 430 2.0 Array | 1449264_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1456464_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1429729_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 229521_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 229521_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300017670.mm.1 | NM_018804.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300017670.mm.1 | NM_018804.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300017670.mm.1 | TC0300002324.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300017671.mm.1 | NM_018804.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300017671.mm.1 | NM_018804.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300017671.mm.1 | TC0300002324.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300017673.mm.1 | NM_018804.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300017673.mm.1 | NM_018804.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300017673.mm.1 | TC0300002324.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300017668.mm.1 | NM_018804.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300017668.mm.1 | NM_018804.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300017668.mm.1 | TC0300002324.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300017672.mm.1 | NM_018804.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300017672.mm.1 | NM_018804.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300017672.mm.1 | TC0300002324.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1456464_x_at | - | - |
| Mouse Expression Set 430 | 1455176_PM_a_at | - | - |
| Mouse Expression Set 430 | 1455176_a_at | - | - |
| Mouse Expression Set 430 | 1456464_PM_x_at | - | - |
| Mouse Expression Set 430 | 1449264_PM_at | - | - |
| Mouse Expression Set 430 | 1449264_at | - | - |
| Mouse Expression Set 430 | 1429315_at | - | - |
| Mouse Expression Set 430 | 1429315_PM_at | - | - |
| Mouse Expression Set 430 | 1429314_PM_at | - | - |
| Mouse Expression Set 430 | 1429314_at | - | - |
| Murine Genome U74v2 Set | 98339_at | - | - |
| Murine Genome U74v2 Set | 98339_PM_at | - | - |
| Murine Genome U74v2 Set | 166580_r_at | - | - |
| Murine Genome U74v2 Set | 165460_at | - | - |
| Murine Genome U74v2 Set | 168346_r_at | - | - |
| Murine Genome U74v2 Set | 166580_PM_r_at | - | - |
| Murine Genome U74v2 Set | 168346_PM_r_at | - | - |
| Murine Genome U74v2 Set | 165460_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1456464_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1456464_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449264_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455176_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449264_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455176_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449264_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1456464_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1456464_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455176_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449264_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455176_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429314_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429314_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429315_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429315_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 100580239 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 630433 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 100580239 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000630433 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1233402 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2431352 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1233402 | - | - |
| mouse-6 v1.1 (Illumina) | scl21963.5_3-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0074029.1_132-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 630433 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 100580239 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl21963.5_3-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 630433 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 100580239 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3141334 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3063644 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1233402 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3141334 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1233402 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3063644 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl21963.5_3-S | - | - |

Syt11
Genomic Map