Assay Details
| Gene Symbol: Acvr2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007396.4 | NP_031422.3 | 6 - 7 | 1483 | 112 | |
| GenBank mRNA | AK041246.1 | - | 6 - 7 | 1436 | 112 | |
| AK143186.1 | - | 7 - 8 | 1582 | 112 | ||
| BC138823.1 | - | 6 - 7 | 1144 | 8861845 | 112 | |
| M65287.1 | - | 6 - 7 | 890 | 112 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Acvr2a
Genomic Map