Assay Details
| Gene Symbol: Adcyap1r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025372.2 | NP_001020543.1 | 2 - 3 | 519 | 96 | |
| NM_007407.4 | NP_031433.3 | 2 - 3 | 519 | 96 | ||
| GenBank mRNA | AK042820.1 | - | 2 - 3 | 511 | 96 | |
| AK083108.1 | - | 2 - 3 | 517 | 96 | ||
| AK147291.1 | - | 2 - 3 | 470 | 96 | ||
| AK147609.1 | - | 2 - 3 | 260 | 96 | ||
| BC067039.1 | - | 2 - 3 | 452 | 6852762 | 96 | |
| D82935.1 | - | 2 - 3 | 202 | 96 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK042820.1,AK147291.1,AK147609.1,BC067039.1,D82935.1,NM_001025372.2,NM_007407.4 transcripts.
Set Membership: |
|

Adcyap1r1
Genomic Map