Assay Details
| Gene Symbol: Adcyap1r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025372.2 | NP_001020543.1 | 5 - 6 | 754 | 78 | |
| NM_007407.4 | NP_031433.3 | 5 - 6 | 754 | 78 | ||
| GenBank mRNA | AK042820.1 | - | 5 - 6 | 746 | 78 | |
| AK047304.1 | - | 4 - 5 | 257 | 78 | ||
| AK081121.1 | - | 4 - 5 | 257 | 78 | ||
| AK083108.1 | - | 5 - 6 | 752 | 78 | ||
| AK147291.1 | - | 5 - 6 | 705 | 78 | ||
| AK147609.1 | - | 5 - 6 | 495 | 78 | ||
| BC067039.1 | - | 5 - 6 | 687 | 6852762 | 78 | |
| D82935.1 | - | 5 - 6 | 437 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adcyap1r1
Genomic Map