Assay Details
| Gene Symbol: Adcyap1r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025372.2 | NP_001020543.1 | 14 - 15 | 1644 | 80 | |
| NM_007407.4 | NP_031433.3 | 15 - 16 | 1728 | 80 | ||
| GenBank mRNA | AK042820.1 | - | 15 - 16 | 1720 | 80 | |
| AK047304.1 | - | 13 - 14 | 1147 | 80 | ||
| AK081121.1 | - | 13 - 14 | 1147 | 80 | ||
| AK083108.1 | - | 15 - 16 | 1727 | 80 | ||
| AK147291.1 | - | 14 - 15 | 1595 | 80 | ||
| AK147609.1 | - | 14 - 15 | 1385 | 80 | ||
| BC067039.1 | - | 14 - 15 | 1577 | 6852762 | 80 | |
| D82935.1 | - | 15 - 16 | 1411 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adcyap1r1
Genomic Map