Assay Details
| Gene Symbol: App | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198823.1 | NP_001185752.1 | 16 - 17 | 2215 | 64 | |
| NM_001198824.1 | NP_001185753.1 | 15 - 16 | 2158 | 64 | ||
| NM_001198825.1 | NP_001185754.1 | 14 - 15 | 2104 | 64 | ||
| NM_001198826.1 | NP_001185755.1 | 15 - 16 | 2161 | 64 | ||
| NM_007471.3 | NP_031497.2 | 14 - 15 | 1990 | 64 | ||
| GenBank mRNA | AK052448.1 | - | 4 - 5 | 411 | 64 | |
| AK076506.1 | - | 8 - 9 | 910 | 64 | ||
| AK128971.1 | - | 14 - 15 | 2113 | 64 | ||
| AK133639.1 | - | 15 - 16 | 2155 | 64 | ||
| AK147543.1 | - | 14 - 15 | 2026 | 64 | ||
| AK148439.1 | - | 15 - 16 | 2034 | 64 | ||
| AK159245.1 | - | 15 - 16 | 2170 | 64 | ||
| AK159718.1 | - | 15 - 16 | 2147 | 64 | ||
| AK161854.1 | - | 9 - 10 | 1071 | 64 | ||
| AK161976.1 | - | 14 - 15 | 1965 | 64 | ||
| AK202486.1 | - | 3 - 4 | 357 | 64 | ||
| AY267348.1 | - | 16 - 17 | 2066 | 64 | ||
| BC005490.1 | - | 12 - 13 | 1579 | 64 | ||
| BC070409.1 | - | 14 - 15 | 1986 | 5708093 | 64 | |
| M18373.1 | - | 14 - 15 | 1979 | 64 | ||
| U84012.1 | - | 14 - 15 | 1979 | 64 | ||
| X59379.1 | - | 14 - 15 | 1909 | 64 | ||
More Information
Set Membership: |
|

App
Genomic Map