Assay Details
| Gene Symbol: Aqp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007472.2 | NP_031498.1 | 1 - 2 | 581 | 64 | |
| GenBank mRNA | AK081886.1 | - | 1 - 2 | 449 | 64 | |
| AK086688.1 | - | 1 - 2 | 449 | 64 | ||
| AK128905.1 | - | 1 - 2 | 448 | 64 | ||
| AK157333.1 | - | 1 - 2 | 449 | 64 | ||
| AK158226.1 | - | 1 - 2 | 449 | 64 | ||
| AK158389.1 | - | 1 - 2 | 449 | 64 | ||
| AK171627.1 | - | 1 - 2 | 449 | 64 | ||
| AK172361.1 | - | 1 - 2 | 449 | 64 | ||
| L02914.1 | - | 1 - 2 | 448 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aqp1
Genomic Map