Assay Details
| Gene Symbol: Aqp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007473.4 | NP_031499.1 | 2 - 3 | 402 | 88 | |
| GenBank mRNA | AB010100.1 | - | 2 - 3 | 293 | 88 | |
| AB056091.1 | - | 2 - 3 | 236 | 88 | ||
| AK076642.1 | - | 2 - 3 | 370 | 88 | ||
| AK131744.1 | - | 2 - 3 | 370 | 88 | ||
| BU945845.1 | - | 2 - 3 | 405 | 88 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AB010100.1,AB056091.1,AK076642.1,AK131744.1,NM_007473.4 transcripts.
Set Membership: |
|

Aqp7
Genomic Map