Assay Details
| Gene Symbol: Slc7a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301424.1 | NP_001288353.1 | 2 - 3 | 154 | 72 | |
| NM_007513.4 | NP_031539.3 | 2 - 3 | 270 | 72 | ||
| GenBank mRNA | AK139342.1 | - | 2 - 3 | 246 | 72 | |
| AK147991.1 | - | 2 - 3 | 179 | 72 | ||
| AK163202.1 | - | 2 - 3 | 154 | 72 | ||
| BC068234.1 | - | 2 - 3 | 162 | 72 | ||
| BC145779.1 | - | 1 - 2 | 75 | 40131082 | 72 | |
| BC145781.1 | - | 1 - 2 | 75 | 40131084 | 72 | |
| M26687.1 | - | 2 - 3 | 188 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK139342.1, AK147991.1, AK163202.1, BC145779.1, BC145781.1, M26687.1, NM_001301424.1, NM_007513.4 transcripts.
Set Membership: |
|

Slc7a1
Genomic Map