Assay Details
| Gene Symbol: Slc7a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301424.1 | NP_001288353.1 | 8 - 9 | 1335 | 74 | |
| NM_007513.4 | NP_031539.3 | 8 - 9 | 1451 | 74 | ||
| GenBank mRNA | AK139342.1 | - | 8 - 9 | 1427 | 74 | |
| AK147991.1 | - | 8 - 9 | 1360 | 74 | ||
| AK163202.1 | - | 8 - 9 | 1335 | 74 | ||
| BC068234.1 | - | 8 - 9 | 1342 | 74 | ||
| BC145779.1 | - | 7 - 8 | 1256 | 40131082 | 74 | |
| BC145781.1 | - | 7 - 8 | 1256 | 40131084 | 74 | |
| JN226407.1 | - | 6 - 7 | 1171 | 74 | ||
| JN226408.1 | - | 6 - 7 | 1171 | 74 | ||
| M26687.1 | - | 8 - 9 | 1369 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc7a1
Genomic Map