Assay Details
| Gene Symbol: Bad | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007522.3 | NP_031548.1 | 1 - 2 | 580 | 66 | |
| GenBank mRNA | BC006762.1 | - | 1 - 2 | 548 | 3257589 | 66 |
| L37296.1 | - | 1 - 2 | 588 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bad
Genomic Map