Assay Details
| Gene Symbol: Bdkrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007539.2 | NP_031565.1 | 1 | 142 | 104 | |
| GenBank mRNA | BC120684.1 | - | 1 | 104 | 40129607 | 104 |
More Information
Updated Mapping Information
Set Membership: |
|

Bdkrb1
Genomic Map