Assay Details
| Gene Symbol: Bmpr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007561.4 | NP_031587.1 | 10 - 11 | 1785 | 81 | |
| GenBank mRNA | AF003942.1 | - | 10 - 11 | 1785 | 81 | |
| AK148180.1 | - | 5 - 6 | 603 | 81 | ||
| AK149385.1 | - | 2666 | 81 | |||
| U78048.1 | - | 10 - 11 | 1427 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bmpr2
Genomic Map