Assay Details
| Gene Symbol: Cacnb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044741.2 | NP_001038206.1 | 5 - 6 | 621 | 75 | |
| NM_007581.3 | NP_031607.2 | 5 - 6 | 875 | 75 | ||
| GenBank mRNA | AF498245.1 | - | 5 - 6 | 473 | 75 | |
| AF498246.1 | - | 5 - 6 | 474 | 75 | ||
| AK190319.1 | - | 2 - 3 | 92 | 75 | ||
| BQ042307.1 | - | 3 - 4 | 294 | 75 | ||
| BU709207.1 | - | 5 - 6 | 466 | 75 | ||
| U20372.1 | - | 5 - 6 | 529 | 75 | ||
More Information
Set Membership: |
|

Cacnb3
Genomic Map