Assay Details
| Gene Symbol: Cacnb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044741.2 | NP_001038206.1 | 6 - 7 | 642 | 66 | |
| NM_007581.3 | NP_031607.2 | 6 - 7 | 896 | 66 | ||
| GenBank mRNA | AF498245.1 | - | 6 - 7 | 494 | 66 | |
| AF498246.1 | - | 6 - 7 | 495 | 66 | ||
| AK190319.1 | - | 3 - 4 | 113 | 66 | ||
| BQ042307.1 | - | 4 - 5 | 315 | 66 | ||
| BU709207.1 | - | 6 - 7 | 487 | 66 | ||
| U20372.1 | - | 6 - 7 | 550 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cacnb3
Genomic Map