Assay Details
| Gene Symbol: Cacnb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044741.2 | NP_001038206.1 | 11 - 12 | 1142 | 73 | |
| NM_001286226.1 | NP_001273155.1 | 10 - 11 | 1315 | 73 | ||
| NM_007581.3 | NP_031607.2 | 11 - 12 | 1396 | 73 | ||
| GenBank mRNA | AF498245.1 | - | 11 - 12 | 994 | 73 | |
| AF498246.1 | - | 11 - 12 | 995 | 73 | ||
| AK148088.1 | - | 10 - 11 | 1316 | 73 | ||
| BC055926.1 | - | 7 - 8 | 1295 | 73 | ||
| BG085743.2 | - | 3 - 4 | 346 | 73 | ||
| CA450725.1 | - | 1 - 2 | 107 | 73 | ||
| U20372.1 | - | 11 - 12 | 73 | |||
More Information
Set Membership: |
|

Cacnb3
Genomic Map