Assay Details
| Gene Symbol: Calcr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042725.1 | NP_001036190.1 | 4 - 5 | 408 | 102 | |
| NM_007588.2 | NP_031614.2 | 4 - 5 | 657 | 102 | ||
| GenBank mRNA | AK039161.1 | - | 4 - 5 | 602 | 102 | |
| AK133983.1 | - | 4 - 5 | 409 | 102 | ||
| AK138275.1 | - | 4 - 5 | 659 | 102 | ||
| AK162391.1 | - | 4 - 5 | 412 | 102 | ||
| BC119232.1 | - | 2 - 3 | 164 | 8733981 | 102 | |
| BC119272.1 | - | 2 - 3 | 164 | 8734021 | 102 | |
| U18542.1 | - | 4 - 5 | 559 | 102 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the U18542.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Citations (1)
Amylin inhibits bone resorption while the calcitonin receptor controls bone formation in vivo.
- Authors
- Dacquin R, Davey RA, Laplace C, Levasseur R, Morris HA, Goldring SR, Gebre-Medhin S, Galson DL, Zajac JD, Karsenty G
- Journal
- J Cell Biol - Page No 509 (Mon Feb 16 00:00:00 EST 2004)
- Volume (Year)
- 164 (2004)
- Product Usage
- -

Calcr
Genomic Map