Assay Details
| Gene Symbol: Ccnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007631.2 | NP_031657.1 | 4 - 5 | 957 | 81 | |
| GenBank mRNA | AK005352.1 | - | 4 - 5 | 651 | 81 | |
| AK053348.1 | - | 4 - 5 | 865 | 81 | ||
| AK149905.1 | - | 4 - 5 | 864 | 81 | ||
| AK164997.1 | - | 4 - 5 | 902 | 81 | ||
| AK170833.1 | - | 4 - 5 | 838 | 81 | ||
| BC044841.1 | - | 4 - 5 | 860 | 3155470 | 81 | |
| BC060172.1 | - | 4 - 5 | 850 | 81 | ||
| M64403.1 | - | 4 - 5 | 912 | 81 | ||
| S78355.1 | - | 4 - 5 | 862 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccnd1
Genomic Map