Assay Details
| Gene Symbol: Ccne1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007633.2 | NP_031659.2 | 2 - 3 | 215 | 84 | |
| GenBank mRNA | AK089950.1 | - | 2 - 3 | 147 | 84 | |
| AK163046.1 | - | 2 - 3 | 204 | 84 | ||
| AK165658.1 | - | 2 - 3 | 205 | 84 | ||
| BC062152.1 | - | 2 - 3 | 242 | 6510163 | 84 | |
| BC084588.1 | - | 2 - 3 | 6506897 | 84 | ||
| BC106191.1 | - | 2 - 3 | 215 | 6336006 | 84 | |
| BC138660.1 | - | 1 - 2 | 48 | 8861680 | 84 | |
| BC138662.1 | - | 1 - 2 | 48 | 8861683 | 84 | |
| X75888.1 | - | 1 - 2 | 55 | 84 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC062152.1,BC084588.1,BC106191.1,BC138660.1,BC138662.1,NM_007633.2,X75888.1 transcripts.
Set Membership: |
|

Ccne1
Genomic Map