Assay Details
| Gene Symbol: Ccne1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007633.2 | NP_031659.2 | 3 - 4 | 296 | 73 | |
| GenBank mRNA | AK089950.1 | - | 3 - 4 | 228 | 73 | |
| AK163046.1 | - | 3 - 4 | 285 | 73 | ||
| AK165658.1 | - | 3 - 4 | 286 | 73 | ||
| BC062152.1 | - | 3 - 4 | 323 | 6510163 | 73 | |
| BC084588.1 | - | 3 - 4 | 340 | 6506897 | 73 | |
| BC106191.1 | - | 3 - 4 | 296 | 6336006 | 73 | |
| BC138660.1 | - | 2 - 3 | 129 | 8861680 | 73 | |
| BC138662.1 | - | 2 - 3 | 129 | 8861683 | 73 | |
| X75888.1 | - | 2 - 3 | 136 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccne1
Genomic Map