Assay Details
| Gene Symbol: Cdk11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007661.3 | NP_031687.2 | 13 - 14 | 1558 | 87 | |
| GenBank mRNA | AK077668.1 | - | 13 - 14 | 1559 | 87 | |
| AK147133.1 | - | 13 - 14 | 1536 | 87 | ||
| BC025058.1 | - | 2 - 3 | 208 | 87 | ||
| BC052920.1 | - | 13 - 14 | 1636 | 6489832 | 87 | |
| L37092.1 | - | 12 - 13 | 2130 | 87 | ||
| M58633.1 | - | 8 - 9 | 901 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdk11b
Genomic Map