Assay Details
| Gene Symbol: Cdx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007673.3 | NP_031699.2 | 1 - 2 | 809 | 88 | |
| GenBank mRNA | AK049566.1 | - | 1 - 2 | 810 | 88 | |
| AK049574.1 | - | 1 - 2 | 681 | 88 | ||
| BC103516.1 | - | 1 - 2 | 687 | 40039012 | 88 | |
| BC103517.1 | - | 1 - 2 | 690 | 40039015 | 88 | |
| BC103519.1 | - | 1 - 2 | 690 | 40039019 | 88 | |
| BC103520.1 | - | 1 - 2 | 690 | 40039017 | 88 | |
| S74520.1 | - | 1 - 2 | 804 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdx2
Genomic Map