Assay Details
| Gene Symbol: Ephb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146351.1 | NP_001139823.1 | 12 - 13 | 2526 | 79 | |
| NM_007680.4 | NP_031706.3 | 12 - 13 | 2522 | 79 | ||
| GenBank mRNA | AK076442.1 | - | 12 - 13 | 2349 | 79 | |
| AK087423.1 | - | 12 - 13 | 2437 | 79 | ||
| AK163823.1 | - | 3 - 4 | 244 | 79 | ||
| BC031924.1 | - | 11 - 12 | 2134 | 4911959 | 79 | |
| L77867.1 | - | 12 - 13 | 2216 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ephb6
Genomic Map