Assay Details
| Gene Symbol: Ephb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146351.1 | NP_001139823.1 | 16 - 17 | 3297 | 82 | |
| NM_007680.4 | NP_031706.3 | 16 - 17 | 3293 | 82 | ||
| GenBank mRNA | AK076442.1 | - | 16 - 17 | 3120 | 82 | |
| AK087423.1 | - | 16 - 17 | 3208 | 82 | ||
| AK163823.1 | - | 7 - 8 | 1015 | 82 | ||
| BC031924.1 | - | 15 - 16 | 2905 | 4911959 | 82 | |
| L77867.1 | - | 16 - 17 | 2987 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ephb6
Genomic Map