Assay Details
| Gene Symbol: Ckm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007710.2 | NP_031736.1 | 6 - 7 | 848 | 64 | |
| GenBank mRNA | AK009950.1 | - | 6 - 7 | 849 | 64 | |
| AK137460.1 | - | 850 | 64 | |||
| BC132424.1 | - | 6 - 7 | 846 | 40130701 | 64 | |
| BC132426.1 | - | 6 - 7 | 846 | 40130703 | 64 | |
| X03233.1 | - | 6 - 7 | 848 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ckm
Genomic Map