Assay Details
| Gene Symbol: Clk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163432.1 | NP_001156904.1 | 9 - 10 | 1352 | 98 | |
| NM_007712.3 | NP_031738.2 | 9 - 10 | 1355 | 98 | ||
| GenBank mRNA | AF033564.1 | - | 9 - 10 | 1077 | 98 | |
| AK134849.1 | - | 3 - 4 | 1596 | 98 | ||
| AK148377.1 | - | 7 - 8 | 2383 | 98 | ||
| AK161259.1 | - | 5 - 6 | 1527 | 98 | ||
| BC015080.1 | - | 9 - 10 | 1340 | 3995512 | 98 | |
| CN538691.1 | - | 4 - 5 | 517 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Clk2
Genomic Map