Assay Details
| Gene Symbol: Clk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF033566.1 | - | 6 - 7 | 737 | 65 | |
| AK013974.2 | - | 4 - 5 | 492 | 65 | ||
| AK161739.1 | - | 2 - 3 | 1746 | 65 | ||
| AY653308.1 | - | 5 - 6 | 627 | 65 | ||
| BC002220.1 | - | 2 - 3 | 293 | 3490616 | 65 | |
| BC012675.1 | - | 6 - 7 | 759 | 3982920 | 65 | |
| CT010293.1 | - | 5 - 6 | 659 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Clk4
Genomic Map