Assay Details
| Gene Symbol: Clk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF033566.1 | - | 9 - 10 | 1126 | 71 | |
| AY653308.1 | - | 8 - 9 | 1016 | 71 | ||
| BC002220.1 | - | 5 - 6 | 682 | 3490616 | 71 | |
| BC012675.1 | - | 9 - 10 | 1148 | 3982920 | 71 | |
| CT010293.1 | - | 8 - 9 | 1048 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Clk4
Genomic Map