Assay Details
| Gene Symbol: Clk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF033566.1 | - | 11 - 12 | 1291 | 68 | |
| AK013974.2 | - | 9 - 10 | 1046 | 68 | ||
| BC002220.1 | - | 7 - 8 | 847 | 3490616 | 68 | |
| BC012675.1 | - | 11 - 12 | 1313 | 3982920 | 68 | |
| CT010293.1 | - | 10 - 11 | 1213 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Clk4
Genomic Map