Assay Details
| Gene Symbol: Csf1r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037859.2 | NP_001032948.2 | 13 - 14 | 2136 | 57 | |
| GenBank mRNA | AK004947.1 | - | 12 - 13 | 1946 | 57 | |
| AK041299.1 | - | 12 - 13 | 1924 | 57 | ||
| AK079247.1 | - | 12 - 13 | 1953 | 57 | ||
| AK080326.1 | - | 12 - 13 | 1934 | 57 | ||
| AK143545.1 | - | 12 - 13 | 1937 | 57 | ||
| AK146069.1 | - | 13 - 14 | 2137 | 57 | ||
| AK154128.1 | - | 12 - 13 | 1951 | 57 | ||
| AK154556.1 | - | 12 - 13 | 1947 | 57 | ||
| AK154653.1 | - | 12 - 13 | 1947 | 57 | ||
| AK154699.1 | - | 12 - 13 | 1947 | 57 | ||
| AK155488.1 | - | 12 - 13 | 1957 | 57 | ||
| AK155576.1 | - | 1953 | 57 | |||
| AK155640.1 | - | 12 - 13 | 1951 | 57 | ||
| AK155654.1 | - | 12 - 13 | 1951 | 57 | ||
| AK155858.1 | - | 12 - 13 | 1954 | 57 | ||
| AK156550.1 | - | 12 - 13 | 1971 | 57 | ||
| AK171386.1 | - | 12 - 13 | 1951 | 57 | ||
| BC036343.1 | - | 12 - 13 | 1923 | 4948135 | 57 | |
| BC043054.1 | - | 12 - 13 | 1927 | 6418718 | 57 | |
| BC050024.1 | - | 12 - 13 | 2034 | 57 | ||
| BC066863.1 | - | 13 - 14 | 2045 | 30436119 | 57 | |
| X06368.1 | - | 12 - 13 | 1929 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Csf1r
Genomic Map