Assay Details
| Gene Symbol: Csk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK013057.1 | - | 3 - 4 | 379 | 67 | |
| AK136876.1 | - | 11 - 12 | 1805 | 67 | ||
| AK137297.1 | - | 11 - 12 | 1218 | 67 | ||
| AK156058.1 | - | 11 - 12 | 1356 | 67 | ||
| AK170815.1 | - | 11 - 12 | 1372 | 67 | ||
| AK190712.1 | - | 1 - 2 | 106 | 67 | ||
| BC018394.1 | - | 11 - 12 | 1331 | 4206879 | 67 | |
| BC052006.1 | - | 11 - 12 | 1389 | 5685297 | 67 | |
| BC095962.1 | - | 13 - 14 | 1712 | 67 | ||
| U05247.1 | - | 9 - 10 | 1083 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Csk
Genomic Map