Assay Details
| Gene Symbol: Dll3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007866.2 | NP_031892.2 | 6 - 7 | 1129 | 90 | |
| GenBank mRNA | AB013440.1 | - | 6 - 7 | 1116 | 90 | |
| AK144282.1 | - | 6 - 7 | 1130 | 90 | ||
| BC052002.1 | - | 6 - 7 | 1106 | 6404029 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dll3
Genomic Map