Assay Details
| Gene Symbol: Dvl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007888.3 | NP_031914.3 | 6 - 7 | 957 | 97 | |
| GenBank mRNA | AK050614.1 | - | 6 - 7 | 761 | 97 | |
| AK144001.1 | - | 6 - 7 | 988 | 98 | ||
| AK146822.1 | - | 6 - 7 | 957 | 97 | ||
| AK159895.1 | - | 6 - 7 | 790 | 97 | ||
| AK168376.1 | - | 6 - 7 | 865 | 97 | ||
| BC053050.1 | - | 6 - 7 | 871 | 6402000 | 97 | |
| BC092396.1 | - | 964 | 30363483 | 97 | ||
| U24160.1 | - | 6 - 7 | 969 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dvl2
Genomic Map