Assay Details
| Gene Symbol: Dvl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007888.3 | NP_031914.3 | 11 - 12 | 1439 | 103 | |
| GenBank mRNA | AK144001.1 | - | 11 - 12 | 1470 | 103 | |
| AK146822.1 | - | 11 - 12 | 1439 | 103 | ||
| AK159895.1 | - | 11 - 12 | 1272 | 103 | ||
| AK168376.1 | - | 11 - 12 | 1347 | 103 | ||
| BC053050.1 | - | 11 - 12 | 1353 | 6402000 | 103 | |
| BC092396.1 | - | 1446 | 30363483 | 103 | ||
| U24160.1 | - | 11 - 12 | 1451 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dvl2
Genomic Map