Assay Details
| Gene Symbol: Dvl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007888.3 | NP_031914.3 | 12 - 13 | 1572 | 87 | |
| GenBank mRNA | AK144001.1 | - | 12 - 13 | 1603 | 87 | |
| AK146822.1 | - | 12 - 13 | 1572 | 87 | ||
| AK159895.1 | - | 12 - 13 | 1405 | 87 | ||
| AK168376.1 | - | 12 - 13 | 1480 | 87 | ||
| BC053050.1 | - | 12 - 13 | 1486 | 6402000 | 87 | |
| BC092396.1 | - | 1579 | 30363483 | 87 | ||
| U24160.1 | - | 12 - 13 | 1584 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dvl2
Genomic Map