Assay Details
| Gene Symbol: Dvl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007889.2 | NP_031915.2 | 3 - 4 | 483 | 89 | |
| GenBank mRNA | AK052151.1 | - | 3 - 4 | 484 | 89 | |
| AK199514.1 | - | 1 - 2 | 92 | 89 | ||
| BC033414.1 | - | 2 - 3 | 1445 | 89 | ||
| U41285.1 | - | 3 - 4 | 565 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dvl3
Genomic Map