Assay Details
| Gene Symbol: Dvl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007889.2 | NP_031915.2 | 10 - 11 | 1184 | 57 | |
| GenBank mRNA | AK052151.1 | - | 10 - 11 | 1185 | 57 | |
| U41285.1 | - | 10 - 11 | 1266 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dvl3
Genomic Map