Assay Details
| Gene Symbol: Ednrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136061.2 | NP_001129533.1 | 5 - 6 | 1360 | 73 | |
| NM_001276296.1 | NP_001263225.1 | 4 - 5 | 1174 | 73 | ||
| NM_007904.4 | NP_031930.1 | 4 - 5 | 1174 | 73 | ||
| GenBank mRNA | AK076426.1 | - | 4 - 5 | 1168 | 73 | |
| AK082103.1 | - | 5 - 6 | 1161 | 73 | ||
| AK083415.1 | - | 5 - 6 | 1057 | 73 | ||
| AK085165.1 | - | 4 - 5 | 1166 | 73 | ||
| AK085532.1 | - | 4 - 5 | 1170 | 73 | ||
| AK198545.1 | - | 1 - 2 | 35 | 73 | ||
| BC026553.1 | - | 5 - 6 | 1286 | 4971909 | 73 | |
| U32329.1 | - | 4 - 5 | 1152 | 73 | ||
| Gene Symbol: Pkm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK197420.1 | - | 73 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Ednrb
Genomic Map