| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1900009741.mm.1 |
NM_001080388.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009741.mm.1 |
NM_001080389.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009741.mm.1 |
NM_001080390.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009741.mm.1 |
NM_007928.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009741.mm.1 |
TC1900001039.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900009746.mm.1 |
NM_001080388.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009746.mm.1 |
NM_001080389.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009746.mm.1 |
NM_007928.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009746.mm.1 |
TC1900001039.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900009747.mm.1 |
NM_001080388.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009747.mm.1 |
NM_001080389.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009747.mm.1 |
NM_007928.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009747.mm.1 |
TC1900001039.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900009753.mm.1 |
NM_001080388.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009753.mm.1 |
NM_001080389.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009753.mm.1 |
NM_007928.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009753.mm.1 |
TC1900001039.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900009756.mm.1 |
NM_001080388.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009756.mm.1 |
NM_001080389.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009756.mm.1 |
NM_007928.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009756.mm.1 |
TC1900001039.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900009757.mm.1 |
NM_001080388.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009757.mm.1 |
NM_001080389.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009757.mm.1 |
NM_007928.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009757.mm.1 |
TC1900001039.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900009758.mm.1 |
NM_001080388.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009758.mm.1 |
NM_001080389.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009758.mm.1 |
NM_007928.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009758.mm.1 |
TC1900001039.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900009751.mm.1 |
NM_001080388.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009751.mm.1 |
NM_001080389.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009751.mm.1 |
NM_007928.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009751.mm.1 |
TC1900001039.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900009744.mm.1 |
NM_001080388.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009744.mm.1 |
NM_001080389.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009744.mm.1 |
NM_007928.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009744.mm.1 |
TC1900001039.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900009745.mm.1 |
NM_001080388.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009745.mm.1 |
NM_001080389.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009745.mm.1 |
NM_007928.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009745.mm.1 |
TC1900001039.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900009748.mm.1 |
NM_001080388.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009748.mm.1 |
NM_001080389.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009748.mm.1 |
NM_007928.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009748.mm.1 |
TC1900001039.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900009749.mm.1 |
NM_001080388.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009749.mm.1 |
NM_001080389.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009749.mm.1 |
NM_007928.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009749.mm.1 |
TC1900001039.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900009752.mm.1 |
NM_001080388.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009752.mm.1 |
NM_001080389.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009752.mm.1 |
NM_007928.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009752.mm.1 |
TC1900001039.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900009762.mm.1 |
NM_001080388.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009762.mm.1 |
NM_001080389.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009762.mm.1 |
NM_007928.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900009762.mm.1 |
TC1900001039.mm.1 |
MTA TC |