Assay Details
| Gene Symbol: Epha4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007936.3 | NP_031962.2 | 13 - 14 | 2403 | 62 | |
| GenBank mRNA | AK028509.1 | - | 13 - 14 | 2396 | 62 | |
| AK147698.1 | - | 13 - 14 | 2399 | 62 | ||
| BC004782.1 | - | 13 - 14 | 2318 | 3588285 | 62 | |
| BC052164.1 | - | 13 - 14 | 2375 | 6512978 | 62 | |
| S57168.1 | - | 13 - 14 | 2403 | 62 | ||
| X57241.1 | - | 1 - 2 | 106 | 62 | ||
| X65138.1 | - | 13 - 14 | 2403 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Epha4
Genomic Map