Assay Details
| Gene Symbol: Epha4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007936.3 | NP_031962.2 | 15 - 16 | 2743 | 70 | |
| GenBank mRNA | AK028509.1 | - | 15 - 16 | 2736 | 70 | |
| AK147698.1 | - | 15 - 16 | 2739 | 70 | ||
| BC004782.1 | - | 15 - 16 | 2658 | 3588285 | 70 | |
| BC052164.1 | - | 15 - 16 | 2715 | 6512978 | 70 | |
| S57168.1 | - | 15 - 16 | 2743 | 70 | ||
| X65138.1 | - | 15 - 16 | 2743 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Epha4
Genomic Map