Assay Details
Gene Symbol: Epha8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007939.2 | NP_031965.2 | 8 - 9 | 1761 | 60 | |
GenBank mRNA | AK220393.1 | - | 6 - 7 | 1245 | 60 | |
U72207.1 | - | 8 - 9 | 1770 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
