Assay Details
Gene Symbol: Epha8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK220393.1 | - | 7 - 8 | 1316 | 67 | |
U72207.1 | - | 9 - 10 | 1841 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
