Assay Details
| Gene Symbol: Pdia3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007952.2 | NP_031978.2 | 7 - 8 | 978 | 96 | |
| GenBank mRNA | AK088721.1 | - | 7 - 8 | 938 | 96 | |
| AK132459.1 | - | 7 - 8 | 943 | 96 | ||
| AK133799.1 | - | 7 - 8 | 937 | 96 | ||
| AK169611.1 | - | 7 - 8 | 932 | 96 | ||
| BC003285.1 | - | 7 - 8 | 897 | 3582199 | 96 | |
| BC033439.1 | - | 7 - 8 | 872 | 4016310 | 96 | |
| M73329.1 | - | 7 - 8 | 917 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pdia3
Genomic Map