Assay Details
| Gene Symbol: Esrra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007953.2 | NP_031979.2 | 1 - 2 | 189 | 154 | |
| GenBank mRNA | AK046588.1 | - | 1 - 2 | 149 | 154 | |
| AK156371.1 | - | 1 - 2 | 165 | 154 | ||
| BC034165.1 | - | 1 - 2 | 132 | 154 | ||
| BC039774.1 | - | 1 - 2 | 201 | 154 | ||
| BC138586.1 | - | 1 - 2 | 149 | 8861606 | 154 | |
| BC171958.1 | - | 1 - 2 | 149 | 9054244 | 154 | |
| U85259.1 | - | 1 - 2 | 156 | 154 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK156371.1, BC138586.1, BC171958.1, NM_007953.2, U85259.1 transcripts.
Set Membership: |
|

Esrra
Genomic Map